2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 2 ports, MPC insert connectors
Thermo Scientific™

2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 2 ports, MPC insert connectors

The small- and medium-sized Thermo Scientific™ Labtainer™ BioProcess Containers (BPCs) range in size from 100 mL to 2 L. TheseRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
SH00018I
also known as SH00018-I
250 mL
SH00017I
also known as SH00017-I
100 mL
SH00019I
also known as SH00019-I
500 mL
SH00020I
also known as SH00020-I
1 L
SH00021I
also known as SH00021-I
2 L
Catalog number SH00018I
also known as SH00018-I
Price (INR)
-
Capacity (Metric):
250 mL
The small- and medium-sized Thermo Scientific™ Labtainer™ BioProcess Containers (BPCs) range in size from 100 mL to 2 L. These bags feature ¼-inch ID tubing, making them ideal for smaller flow applications. The MPC insert connectors are easy to use and offer secure fluid handling. The 1 L Thermo Scientific™ BPC 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL ethyl vinyl acetate film, and includes 2 ports (2 MPC insert + barb and cap).

Features of the system include:
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications include:
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquid
• Collecting chromatography feed and fraction
• Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, and steam-to and steam-through connectors
• Filters (used for bioburden reduction)—sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers and platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands

Specifications
Dimensions16.5 cm x 12.4 cm (6.50 in. x 4.88 in.)
Outer PackagingTwo 4-mil. poly bags
Product LineLabtainer
Quantity250 mL, 50 per case
RegulatorycGMP
Shipping ConditionRoom Temperature
Sterilization MethodGamma Irradiated
Capacity (Metric)250 mL
Film DescriptionASI 28
MaterialEVA
Port DetailMPC Insert Connector
Tubing StyleEVA
Unit Size250 mL
Contents & Storage
Store at room temperature.