The small- and medium-sized Thermo Scientific™ Labtainer™ BioProcess Containers (BPCs) range in size from 100 mL to 2 L. TheseRead more
Have Questions?
Change view
Catalog Number
Capacity (Metric)
SH00020I
also known as SH00020-I
1 L
SH00017I
also known as SH00017-I
100 mL
SH00018I
also known as SH00018-I
250 mL
SH00019I
also known as SH00019-I
500 mL
SH00021I
also known as SH00021-I
2 L
5 Options
Catalog number SH00020I
also known as SH00020-I
Price (INR)
-
Capacity (Metric):
1 L
The small- and medium-sized Thermo Scientific™ Labtainer™ BioProcess Containers (BPCs) range in size from 100 mL to 2 L. These bags feature ¼-inch ID tubing, making them ideal for smaller flow applications. The MPC insert connectors are easy to use and offer secure fluid handling. The 1 L Thermo Scientific™ BPC 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL ethyl vinyl acetate film, and includes 2 ports (2 MPC insert + barb and cap).
Features of the system include: • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks • All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions
Applications include: • Harvesting from, feeding into, and collecting samples from bioreactors and fermenters • Preparing and storing buffer, culture media, and process liquid • Collecting chromatography feed and fraction • Harvesting, storing, and transporting bulk drug product and drug precursors
Customize standard BPC systems for optimized single-use technology Choose from the industry’s largest component library, with over 2,000 unique components that include: • Fittings—straight, reducing, elbow, T-style, X-style, and cross-style • Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, and steam-to and steam-through connectors • Filters (used for bioburden reduction)—sterilization grade filters and vent filters • Tubing—thermoplastic elastomers and platinum-cured silicone • Sample ports—septum-style and needle-free ports • Clamps—sliders, pinch, and crimp bands
Specifications
Dimensions28.6 cm x 13.3 cm (11.25 in. X 5.25 in.)