2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 2 ports, Luer Lock body connector and winged spike port
Thermo Scientific™

2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 2 ports, Luer Lock body connector and winged spike port

The 100 mL Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, aRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
SH00027I
also known as SH00027-I
500 mL
SH00025I
also known as SH00025-I
100 mL
SH00026I
also known as SH00026-I
250 mL
SH00028I
also known as SH00028-I
1 L
SH00029I
also known as SH00029-I
2 L
Catalog number SH00027I
also known as SH00027-I
Price (INR)
-
Capacity (Metric):
500 mL
The 100 mL Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 2 ports (1 luer body + cap, 1 winged spike port).

Features of the system include:
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications include:
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquid
• Collecting chromatography feed and fraction
• Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, and steam-to and steam-through connectors
• Filters (used for bioburden reduction)—sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers and platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands

Specifications
Dimensions18.4 cm x 12.4 cm (7.25 in. X 4.88 in.)
Outer PackagingTwo 4-mil. poly bags
Product LineLabtainer
Quantity500 mL, 50 per case
RegulatorycGMP
Shipping ConditionRoom Temperature
Sterilization MethodGamma Irradiated
Capacity (Metric)500 mL
Film DescriptionASI 28
MaterialEVA
Port DetailLuer Lock Body Connector and Winged Spike Port
Tubing StyleEVA
Unit Size500 mL
Contents & Storage
Store at room temperature.