2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 1 port, Luer Lock body connector
2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 1 port, Luer Lock body connector
Thermo Scientific™

2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 1 port, Luer Lock body connector

The 100 mL Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, aRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
SH00037I
also known as SH00037-I
500 mL
SH00035I
also known as SH00035-I
100 mL
SH00036I
also known as SH00036-I
250 mL
SH00038I
also known as SH00038-I
1 L
SH00039I
also known as SH00039-I
2 L
Catalog number SH00037I
also known as SH00037-I
Price (INR)
-
Request A Quote
Capacity (Metric):
500 mL
The 100 mL Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 1 port (1 luer body + cap).

Features of the system include:
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications include:
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquid
• Collecting chromatography feed and fraction
• Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, and steam-to and steam-through connectors
• Filters (used for bioburden reduction)—sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers and platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands

Specifications
Dimensions18.4 cm x 12.4 cm (7.25 in. X 4.88 in.)
Outer PackagingTwo 4-mil. poly bags
Product LineLabtainer
Quantity500 mL, 50 per case
RegulatorycGMP
Shipping ConditionRoom Temperature
Sterilization MethodGamma Irradiated
Capacity (Metric)500 mL
Film DescriptionASI 28
MaterialEVA
Port DetailLuer Lock Body Connector
Tubing StyleEVA
Unit Size500 mL
Contents & Storage
Store at room temperature.