Productainer™ BioProcess Container (BPC), 50 L with 2 Top Ports, 3D Single-Use
Thermo Scientific™

Productainer™ BioProcess Container (BPC), 50 L with 2 Top Ports, 3D Single-Use

The Thermo Scientific™ Productainer™ BioProcess Container (BPC) 50 L 3D system is composed of CX5-14 single web, multi-layer plastic filmRead more
Have Questions?
Catalog NumberQuantity
SH3064901
also known as SH30649.01
50 L
Catalog number SH3064901
also known as SH30649.01
Price (INR)
-
Quantity:
50 L
The Thermo Scientific™ Productainer™ BioProcess Container (BPC) 50 L 3D system is composed of CX5-14 single web, multi-layer plastic film and includes 2 top ports. The 3D square tube design with top dispense is for use with flat-bottom Thermo Scientific™ drums.

Features:
• Use this system to eliminate post-use cleaning steps required with re-useable containers, and to reduce cross-contamination risks.
• All 3D BPCs are constructed in an ISO 7 cleanroom under cGMP conditions.
• All 3D BPCs are designed to fit the full range of support containers, both square and cylindrical – from 50 to 2000 L.

Applications of the Thermo Scientific 3D BPC include:
• Hydration and sterilization of process buffers, liquids, and culture media.
• Chromatography feed and fraction collection.
• Storage and transport of bulk drug product and bulk drug precursors.
• Harvest from and feed into bioreactors and fermentors.
• Dispensing, packaging, and storage of cell culture media, buffers and process liquids.

Customize standard BPC systems for optimized single-use technology. Choose from the industry’s largest component library – with over 20,000 unique components that include:
Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
Filters—used for bioburden reduction; sterilization grade filters and vent filters.
Tubing—thermoplastic elastomers, platinum-cured silicone.
Sample ports—septum-style and needle-free ports.
Clamps—sliders, pinch, and crimp bands.

Specifications
Film DescriptionCX5-14
For Use With (Equipment)SV50076.02
MaterialPolyethylene
Outer PackagingPackages placed in strong cardboard cartons with product and lot identification
Port DetailTop Ports: 2 x 0.38 in.
Product LineProductainer
Protective Inner PackagingPacked and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Quantity50 L
Sterilization MethodGamma Irradiated
Tubing Line 1Line 1. MPC insert, polycarbonate, Tubing: C-Flex, length: 45.7 cm (18 in.) ID x OD: 9.5 x 15.9mm (0.38 x 0.63 in.)
Tubing Line 2Line 2. MPC body, polycarbonate Tubing: C-Flex, length: 45.7 cm (18 in.) ID x OD: 9.5 x 15.9 mm (0.38 x 0.63 in.)
Capacity (Metric)50 L
Unit Size50 L