Productainer™ BioProcess Container (BPC), 200 L with 4 TopPorts and 1 Bottom Dispense, 3D Single-Use BPC
Thermo Scientific™

Productainer™ BioProcess Container (BPC), 200 L with 4 TopPorts and 1 Bottom Dispense, 3D Single-Use BPC

The Thermo Scientific™ Productainer™ BioProcess Container (BPC), 200 L 3D system is composed of CX5-14 single web, multi-layer plastic filmRead more
Have Questions?
Catalog NumberQuantity
SH3065203200 L
Catalog number SH3065203
Price (INR)
-
Quantity:
200 L
The Thermo Scientific™ Productainer™ BioProcess Container (BPC), 200 L 3D system is composed of CX5-14 single web, multi-layer plastic film and includes 4 top ports and 1 bottom port. The 3D square tube design with bottom port dispense is for use with conical, bottom-drain drums.

Features:
• Use this system to eliminate post-use cleaning steps required with re-useable containers, and to reduce cross-contamination risks.
• All 3D BPCs are constructed in an ISO 7 cleanroom under cGMP conditions.
• All 3D BPCs are designed to fit the full range of support containers, both square and cylindrical – from 50 to 2000 L.

Applications of the Thermo Scientific 3D BPC include:
• Hydration and sterilization of process buffers, liquids, and culture media.
• Chromatography feed and fraction collection.
• Storage and transport of bulk drug product and bulk drug precursors.
• Harvest from and feed into bioreactors and fermentors.
• Dispensing, packaging, and storage of cell culture media, buffers and process liquids.

Customize standard BPC systems for optimized single-use technology. Choose from the industry’s largest component library – with over 20,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
• Filters—used for bioburden reduction; sterilization grade filters and vent filters.
• Tubing—thermoplastic elastomers, platinum-cured silicone.
• Sample ports—septum-style and needle-free ports.
• Clamps—sliders, pinch, and crimp bands.

Specifications
For Use With (Equipment)SV50517.06
Outer PackagingPackages placed in strong cardboard cartons with product and lot identification
Product LineProductainer
Protective Inner PackagingPacked and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Quantity200 L
Size200 L dip tube: 81 cm (32 in.)
Sterilization MethodGamma Irradiated
Tubing Line 1Line 1. MPC insert, polycarbonate, Tubing: C-Flex, length: 121.9 cm (48 in.) dip tube ID x OD: 9.5 x 15.9 mm (0.38 in. x 0.63 in.) with dip tube length: 30.5 cm (12 in.) Top Port
Tubing Line 2Line 2. Insert luer lock connection, polypropylene Tubing: C-Flex, length: 121.9 cm (48 in.) ID x OD: 6.4 x 9.5 mm (0.25 in. x 0.38 in.) Top Port
Tubing Line 3Line 3. Body luer lock connection, polypropylene Tubing: C-Flex, length: 121.9 cm (48 in.) ID x OD: 6.4 x 9.5 mm (0.25 in. x 0.38 in.) Top Port
Tubing Line 4Line 4. Plugged, no tubing
Tubing Line 5Line 5. MPC body, polycarbonate Tubing: silicone, length: 121.9 cm (48 in.) ID x OD: 9.5 x 15.9 mm (0.38 in. x 0.63 in.) Bottom port
Capacity (Metric)200 L
Film DescriptionCX5-14
MaterialPolyethylene
Port DetailBottom Port: 1 x 0.38 in., Top Ports: 1 x 0.38 in. 2 x 0.25 in.
Unit Size200 L