Labtainer™ BioProcess Container (BPC), 250mL with 2 Ports, Luer Lock and MPC Insert, 2D BPC
Thermo Scientific™

Labtainer™ BioProcess Container (BPC), 250mL with 2 Ports, Luer Lock and MPC Insert, 2D BPC

Have Questions?
Catalog NumberQuantity
SH3066213
also known as SH30662.13
250 mL
Catalog number SH3066213
also known as SH30662.13
Price (INR)
-
Quantity:
250 mL
The 250 mL Thermo Scientific™ Labtainer™ BPC, 2D system is composed of CX5-14 single-web, multi-layer film, and includes 2 edge ports with luer lock body and MPC insert.

Features:

  • Use this system to eliminate post-use cleaning steps required with re-useable containers, and to reduce cross-contamination risks.
  • All Labtainers are constructed in an ISO 7 cleanroom under cGMP conditions.

Applications of the Thermo Scientific Labtainer BPC include:

  • Harvest from, feed into, and collect samples from bioreactors and fermentors.
  • Buffer, culture media, and process liquid preparation and storage.
  • Chromatography feed and fraction collection.
  • Harvest, storage and transport of bulk drug product and bulk drug precursors.

Customize standard BPC systems for optimized single-use technology

Choose from the industry's largest component library – with over 20,000 unique components that include:

  • Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
  • Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
  • Filters—used for bioburden reduction; sterilization grade filters and vent filters.
  • Tubing—thermoplastic elastomers, platinum-cured silicone.
  • Sample ports—septum-style and needle-free ports.
  • Clamps—sliders, pinch, and crimp bands.
Specifications
Capacity (Metric)250 mL
Dimensions (L x W x H)19.1cm x 15cm (7.5 in. x 5.9 in.)
Film DescriptionCX5-14
Includes2 Edge Port
Length (Metric)19.1 cm
Outer PackagingPackages placed in strong cardboard cartons with product and lot identification
Port DetailEdge Ports: 2 x 0.25 in.
Product LineLabtainer
Protective Inner PackagingPacked and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Quantity250 mL
Sterilization MethodGamma Irradiated
Tubing Line 1Line 1. Body luer lock connection,polypropylene Tubing: C-Flex, length: 30 cm (12 in.) Id x OD: 3.2 x 6.4 mm (0.13 x 0.25 in.)
Tubing Line 2Line 2. MPC insert, polycarbonate, No Tubing
Width (Metric)15 cm
Length (English)7.5 in.
MaterialPlastic Film
Product TypeBioprocess Container for Liquid
Width (English)5.9 in.
Unit Size250 mL