Labtainer™ BioProcess Container (BPC), 20L with 3 Ports, MPC Insert, MPC Body, and Luer Lock, 2D BPC
Thermo Scientific™

Labtainer™ BioProcess Container (BPC), 20L with 3 Ports, MPC Insert, MPC Body, and Luer Lock, 2D BPC

The 20 L Thermo Scientific™ Labtainer™ BPC, 2D system is composed of CX5-14 single web, multi-layer plastic film and includesRead more
Have Questions?
Catalog NumberQuantity
SH307120320 L, each
Catalog number SH3071203
Price (INR)
-
Quantity:
20 L, each
The 20 L Thermo Scientific™ Labtainer™ BPC, 2D system is composed of CX5-14 single web, multi-layer plastic film and includes 3 top ports (MPC insert, MPC body, and luer lock body connection).

Features:
• Use this system to eliminate post-use cleaning steps required with re-useable containers, and to reduce cross-contamination risks.
• All Labtainers are constructed in an ISO 7 cleanroom under cGMP conditions.

Applications of the Thermo Scientific Labtainer BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors.
• Buffer, culture media, and process liquid preparation and storage.
• Chromatography feed and fraction collection.
• Harvest, storage and transport of bulk drug product and bulk drug precursors.

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library – with over 20,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
• Filters—used for bioburden reduction; sterilization grade filters and vent filters.
• Tubing—thermoplastic elastomers, platinum-cured silicone.
• Sample ports—septum-style and needle-free ports.
• Clamps—sliders, pinch, and crimp bands.

Specifications
Dimensions65.5 cm x 43.2 cm (25.8 in. x 17 in.)
Outer PackagingPackages placed in strong cardboard cartons with product and lot identification
Product LineLabtainer
Quantity20 L, each
Sterilization MethodGamma Irradiated
Tubing Line 1Line 1. MPC insert, polycarbonate, Tubing: C-Flex, length: 61 cm (24 in.) ID x OD: 9.5 x 15.9mm (0.38 x 0.63 in.)
Tubing Line 2Line 2. MPC body, polycarbonate, Tubing: C-Flex, length: 61 cm (24 in.) ID x OD: 9.5 x 15.9 mm (0.38 x 0.63 in.)
Tubing Line 3Line 3. Body luer lock connection, polypropylene Tubing: C-Flex, length: 61 cm (24 in.) ID x OD: 3.2 x 6.4 mm (0.13 x 0.25 in.)
Capacity (Metric)20 L
Film DescriptionCX5-14
Port DetailEdge Ports: 2 x 0.38 in. 1 x 0.25 in.
Unit Size20 L