HyPerforma™ Smartainer™ 3D BioProcess Container (BPC), 200L
Thermo Scientific™

HyPerforma™ Smartainer™ 3D BioProcess Container (BPC), 200L

The 200 L Thermo Scientific large-volume BPC, 3D system is composed of CX5-14 single web, multi-layer plastic film and includes 2 top ports (2 MPX inserts), 1 bottom drain (MPX insert).
Have Questions?
Catalog NumberQuantity
SH3071702
also known as SH30717.02
200 L
Catalog number SH3071702
also known as SH30717.02
Price (INR)
-
Quantity:
200 L

The 200 L Thermo Scientific™ large-volume BPC, 3D system is composed of CX5-14 single web, multi-layer plastic film and includes 2 top ports (2 MPX inserts), 1 bottom drain (MPX insert).

Features:

  • Use this system to eliminate post-use cleaning steps required with re-useable containers, and to reduce cross-contamination risks.
  • All 3D BPCs are constructed in an ISO 7 cleanroom under cGMP conditions.
  • All 3D BPCs are designed to fit the full range of support containers, both square and cylindrical – from 50 to 2000 L.

Applications of the Thermo Scientific 3D BPC include:

  • Hydration and sterilization of process buffers, liquids, and culture media.
  • Chromatography feed and fraction collection.
  • Storage and transport of bulk drug product and bulk drug precursors.
  • Harvest from and feed into bioreactors and fermentors.
  • Dispensing, packaging, and storage of cell culture media, buffers and process liquids.

Customize standard BPC systems for optimized single-use technology

Choose from the industry's largest component library – with over 20,000 unique components that include:

  • Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
  • Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
  • Filters—used for bioburden reduction; sterilization grade filters and vent filters.
  • Tubing—thermoplastic elastomers, platinum-cured silicone.
  • Sample ports—septum-style and needle-free ports.
  • Clamps—sliders, pinch, and crimp bands.
Specifications
Film DescriptionCX5-14
For Use With (Equipment)Smartainer™ Hardware Stainless Steel Outer Container (Cat. No. SV50160.01)
Outer PackagingPackages placed in strong cardboard cartons with product and lot identification
Port DetailBottom Port 1 x 0.5 in., Top Ports: 2 x 0.5 in.
Product LineHyPerforma™, Smartainer™ II
Protective Inner PackagingPacked and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Quantity200 L
Sterilization MethodGamma Irradiated
Tubing Length Line 151 cm (20 in.)
Tubing Length Line 251 cm (20 in.)
Tubing Length Line 351 cm (20 in.)
Tubing Line 1MPX Body, Polycarbonate, ID x OD: 12.7 x 19.1 mm (0.5 x 0.75 in.) Top Port
Tubing Line 2MPX Body, Polycarbonate, ID x OD: 12.7 x 19.1 mm (0.5 x 0.75 in.) Top Port
Tubing Line 3MPX Body, Polycarbonate, ID x OD: 12.7 x 19.1 mm (0.5 x 0.75 in.) Bottom Port
Tubing Material Line 1Silicone
Tubing Material Line 2Silicone
Tubing Material Line 3Silicone
Capacity (Metric)200 L
Unit Size200 L