Product Image
Thermo Scientific™
Accessories for HyPerforma™ Single-Use Bioreactors, Bioreactor Probe Assembly
The Thermo Scientific™ Bioreactor Probe Assembly (non-sterile) is used to package the pH and DO probes for sterilization and to aseptically connect them to the BPCs for Thermo Scientific™ HyPerforma™ Single-Use Bioreactors.
Have Questions?
Catalog NumberQuantity
SH30720011 each
Catalog number SH3072001
Price (INR)
-
Quantity:
1 each
The Thermo Scientific™ Bioreactor Probe Assembly (non-sterile) is used to package the pH and DO probes for sterilization and to aseptically connect them to the BPCs for Thermo Scientific™ HyPerforma™ Single-Use Bioreactors.
Specifications
For Use With (Application)50 to 1000 L
Product LineHyPerforma
Quantity1 each
SterilityNon-sterile
Sterilization MethodNon-sterile
Unit Size1 each