PowderTainer™ II 1 to 25 kg Single-Use BPC for powder applications, 1 kg
Thermo Scientific™

PowderTainer™ II 1 to 25 kg Single-Use BPC for powder applications, 1 kg

The 1 kg Thermo Scientific™ Powdertainer™ II BioProcess Container (BPC) is a 2D powder handling system composed of CX3-9 film that includes 2 ports (1 wash down line, 1 powder port).
Have Questions?
Catalog NumberCapacity (Metric)
SH3073701
also known as SH30737.01
1 kg
Catalog number SH3073701
also known as SH30737.01
Price (INR)
-
Capacity (Metric):
1 kg

The 1 kg Thermo Scientific™ Powdertainer™ II BioProcess Container (BPC) is a 2D powder handling system composed of CX3-9 film that includes 2 ports (1 wash down line, 1 powder port). The Powdertainer BPC is specifically designed for powder containment and discharge applications, and maintains a closed system for maximum recovery of powder while minimizing the risk of cross contamination.

Features:

  • Three-inch tri-clamp port designed for secure connection to, and easy integration, with existing formulation vessels.
  • Powdertainer BPC features suspensor handle for support during discharge, as well as neck clamp to retain powder prior to discharge.
  • Use this system to eliminate post-use cleaning steps required with re-useable containers.

Applications of the Thermo Scientific Powdertainer BPC include:

  • Storage and delivery of powder culture media and buffers.
  • Storage and dispensing of pre-weighed chemicals and other process powders.
Specifications
Capacity (Metric)1 kg
Dimensions (L x W)16.8 x 12.5 in. (42.7 x 31.8 cm)
Film DescriptionCX3-9
For Use With (Application)Powder Application
Includes2 Ports (1 Wash Down Line, 1 Powder Port)
Port DetailPowder port: 7.6 cm (3 in.) tri-clamp with integrated seal
Product LinePowderTainer™ II
Tubing StyleSilicone
Product TypeBioprocess Container for Powder