Product Image
Thermo Scientific™
BioProcess Containers (BPC), 500L, for HyPerforma™ Single-Use Mixer and docking tank, liquid-to-liquid
The 500L Thermo Scientific™ Bioprocess Container (BPC) is for use with the Thermo Scientific™ HyPerforma™ Single-Use Mixer and docking tankRead more
Have Questions?
Catalog NumberCapacity (Metric)
SH30754.01
also known as SH3075401
500 L
Catalog number SH30754.01
also known as SH3075401
Price (INR)
-
Capacity (Metric):
500 L
The 500L Thermo Scientific™ Bioprocess Container (BPC) is for use with the Thermo Scientific™ HyPerforma™ Single-Use Mixer and docking tank for liquid-to-liquid mixing.
Single Use Mixer (S.U.M.) BPC options include:
• Open top liner for media/buffer hydration
• Closed BPC with powder port for contained media/buffer hydration
• Closed BPC with powder port and monitoring probe capability
• Closed BPC for liquid-liquid mixing of critical sterile solutions such as pooling of fractions or processing of intermediate solutions
• Closed BPC for liquid-liquid mixing with monitoring probe capability
Specifications
Film DescriptionCX5-14
For Use With (Application)Liquid-Liquid BPC
Port Detail0 Probe Port
Product LineHyPerforma
Capacity (Metric)500 L