BioProcess Container (BPC), 50L, for HyPerforma™ Single-Use Mixer, liquid-to-liquid w/probe ports
Thermo Scientific™
BioProcess Container (BPC), 50L, for HyPerforma™ Single-Use Mixer, liquid-to-liquid w/probe ports
Have Questions?
Catalog NumberQuantity
SH30767.0250 L
Catalog number SH30767.02
Price (INR)
-
Quantity:
50 L
The 50L Thermo Scientific™ Bioprocess Container (BPC) for liquid-to-liquid mixing applications, is for use with the Thermo Scientific™ HyPerforma™ Single-Use Mixer and is composed of CX 5-14 single web, multi-layer plastic film and includes probe ports.
Single Use Mixer (S.U.M.) BPC options include:
• Open top liner for media/buffer hydration
• Closed BPC with powder port for contained media/buffer hydration
• Closed BPC with powder port and monitoring probe capability
• Closed BPC for liquid-liquid mixing of critical sterile solutions such as pooling of fractions or processing of intermediate solutions
• Closed BPC for liquid-liquid mixing with monitoring probe capability
Specifications
Product LineHyPerforma
Quantity50 L
Capacity (Metric)50 L
Unit Size50 L