BioProcess Container (BPC), 2000L, for HyPerforma™ Single-Use Mixer, powder-to-liquid
Thermo Scientific™
BioProcess Container (BPC), 2000L, for HyPerforma™ Single-Use Mixer, powder-to-liquid
The 2000L Thermo Scientific™ Bioprocess Container (BPC) for powder-to-liquid mixing applications, is for use with the Thermo Scientific™ HyPerforma™ Single-UseRead more
Have Questions?
Catalog NumberCapacity (Metric)
SH30770.012000 L
Catalog number SH30770.01
Price (INR)
-
Capacity (Metric):
2000 L
The 2000L Thermo Scientific™ Bioprocess Container (BPC) for powder-to-liquid mixing applications, is for use with the Thermo Scientific™ HyPerforma™ Single-Use Mixer and is composed of CX 5-14 single web, multi-layer plastic film with 0 ports.
Single Use Mixer (S.U.M.) BPC options include:
• Open top liner for media/buffer hydration
• Closed BPC with powder port for contained media/buffer hydration
• Closed BPC with powder port and monitoring probe capability
• Closed BPC for liquid-liquid mixing of critical sterile solutions such as pooling of fractions or processing of intermediate solutions
• Closed BPC for liquid-liquid mixing with monitoring probe capability
Specifications
Film DescriptionCX5-14
For Use With (Application)Powder-Liquid BPC
Port Detail0 Probe Port
Product LineHyPerforma
Capacity (Metric)2000 L