Impeller sleeve for open-top mixing, for 2000L HyPerforma™ Single-Use Mixer BioProcess Container (BPC)
Thermo Scientific™
Impeller sleeve for open-top mixing, for 2000L HyPerforma™ Single-Use Mixer BioProcess Container (BPC)
Support mixing applications in bioproduction using Thermo Scientific™ Bioprocess Containers (BPC) for Thermo Scientific™ HyPerforma™ Single-Use Mixer, with impeller-based mixingRead more
Have Questions?
Catalog NumberQuantity
SH30772.01Each
Catalog number SH30772.01
Price (INR)
-
Quantity:
Each
Support mixing applications in bioproduction using Thermo Scientific™ Bioprocess Containers (BPC) for Thermo Scientific™ HyPerforma™ Single-Use Mixer, with impeller-based mixing in open or closed designs.
Single Use Mixer (S.U.M.) BPC options include:
• Open top liner for media/buffer hydration
• Closed BPC with powder port for contained media/buffer hydration
• Closed BPC with powder port and monitoring probe capability
• Closed BPC for liquid-liquid mixing of critical sterile solutions such as pooling of fractions or processing of intermediate solutions
• Closed BPC for liquid-liquid mixing with monitoring probe capability
Specifications
Capacity (Metric)2000 L
Product LineHyPerforma
QuantityEach
SterilitySterile
Product TypeImpeller Sleeve
Unit Size2000 L