Dolly for 50 to 200L Plastic Drums for HyPerforma™ Single-Use Mixer DS 300 (single, polyethylene)
Thermo Scientific™
Dolly for 50 to 200L Plastic Drums for HyPerforma™ Single-Use Mixer DS 300 (single, polyethylene)
The Dolly (single pack) is designed for use with 50 to 200 L Plastic Drums and the Thermo Scientific™ HyPerforma Single-Use Mixer (S.U.M.) DS 300.
Have Questions?
Catalog NumberQuantity
SH3095801
also known as SH30958.01
1 Pk.
Catalog number SH3095801
also known as SH30958.01
Price (INR)
-
Quantity:
1 Pk.
The Dolly (single pack) is designed for use with 50 to 200 L Plastic Drums and the Thermo Scientific™ HyPerforma™ Single-Use Mixer (S.U.M.) DS 300. Dollies for the drums are available in two sizes: one for 50 to 200 L drums and another for 300 L drums. These drums should be used with the corresponding optional drum positioner that is available with the docking station.
Required accessories:
  • HyPerforma Single-Use Mixer DS 300 docking station
  • HyPerforma Single-Use Mixer DS 300 bioprocess container bags for closed-top mixing or liners for open-top mixing
Recommended for:
  • Powder-liquid or liquid-liquid mixing applications in single-use
  • High-throughput mixing and storage
Specifications
MaterialPolyethylene
Product LineHyPerforma
Quantity1 Pk.
Capacity (Metric)50 to 200 L
For Use With (Equipment)DS 300 Single-Use Mixer
Unit Size1 pack