Productainer™ 3D BioProcess Container (BPC), Aegis™ 5-14 film, three ports, tri-clamps
Thermo Scientific™

Productainer™ 3D BioProcess Container (BPC), Aegis™ 5-14 film, three ports, tri-clamps

Thermo Scientific Productainer 3D BioProcess Containers (BPCs) are used to package, store, and transport media, buffers, water for injection (WFI),Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
SH3096803200 L
SH309680150 L
SH3096802100 L
Catalog number SH3096803
Price (INR)
-
Capacity (Metric):
200 L

Thermo Scientific Productainer 3D BioProcess Containers (BPCs) are used to package, store, and transport media, buffers, water for injection (WFI), process liquids, and biological products. Productainer BPCs are constructed with Aegis 5-14 single web, multi-layered cast film manufactured in a cGMP clean room facility. Our standard Productainer BPCs are suitable for a wide array of bioprocess applications. For optimal fit, form, and function, Productainer BPCs can be customized using the industry's largest component library.

Highlights:
• Film optimized for flexibility and durability—superior stress resistance, puncture resistance, and physical barrier properties to ensure stability of contents
• Gamma-irradiation sterilized
• Squared tube design with polyethylene ports located on the top of the BPC for easy access
• Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
• Customizable via the industry's largest library of qualified components

Recommended for:
• Harvest—collection and storage of bulk product from bioreactors
• Storage—sterile storage of media, buffers, and process intermediates
• Separation—feeding and receiving of sterile liquids from separation systems
• Purification—buffer feeding to purification system, fraction collection and storage
• Filling—sterile reservoir for filling systems
• Transport and shipping—bulk sterile liquids, buffers, media and process intermediates within and between facilities

Specifications
Quantity200 L
Capacity (Metric)200 L
Tubing StyleC-Flex
Unit Size200 L