BioProcess Container (BPC), 50L, for HyPerforma™ Single-Use Mixer, powder-to-liquid, Aegis™ 5-14 film, 0 ports
Thermo Scientific™
BioProcess Container (BPC), 50L, for HyPerforma™ Single-Use Mixer, powder-to-liquid, Aegis™ 5-14 film, 0 ports
The 50L Thermo Scientific™ Bioprocess Containers (BPC) for powder-to-liquid mixing, is composed of Aegis 5-14 single web, multi-layer plastic filmRead more
Have Questions?
Catalog NumberQuantity
SH30973.01Each
Catalog number SH30973.01
Price (INR)
-
Quantity:
Each
The 50L Thermo Scientific™ Bioprocess Containers (BPC) for powder-to-liquid mixing, is composed of Aegis 5-14 single web, multi-layer plastic film and includes 0 ports.
Single Use Mixer (S.U.M.) BPC options include:
• Open top liner for media/buffer hydration
• Closed BPC with powder port for contained media/buffer hydration
• Closed BPC with powder port and monitoring probe capability
• Closed BPC for liquid-liquid mixing of critical sterile solutions such as pooling of fractions or processing of intermediate solutions
• Closed BPC for liquid-liquid mixing with monitoring probe capability
Specifications
Port Detail0 ports
Product LineHyPerforma
QuantityEach
Capacity (Metric)50 L
Unit Size50 L