Thermo Scientific™ Single-Use Fermentor BioProcess Container, Aegis™ 5-14 film, pH/DO sensor, foam sensor, Meissner inlet, two 5-inch exhaust filters, 30 L
Thermo Scientific™ Single-Use Fermentor BioProcess Container, Aegis™ 5-14 film, pH/DO sensor, foam sensor, Meissner inlet, two 5-inch exhaust filters, 30 L
Thermo Scientific™
Thermo Scientific™ Single-Use Fermentor BioProcess Container, Aegis™ 5-14 film, pH/DO sensor, foam sensor, Meissner inlet, two 5-inch exhaust filters, 30 L
This 30 L Thermo Scientific Single-Use Fermentor (SUF) BioProcess Container (BPC) is composed of Aegis 5-14 film, a five-layer, animal-originRead more
Have Questions?
Catalog NumberQuantity
SH310100230 L
Catalog number SH3101002
Price (INR)
-
Request A Quote
Quantity:
30 L
This 30 L Thermo Scientific Single-Use Fermentor (SUF) BioProcess Container (BPC) is composed of Aegis 5-14 film, a five-layer, animal-origin free film, and includes a Mettler Toledo single-use pH and DO sensor, foam sensor, Meissner inlet, two 5-inch exhaust filters, and condenser.
All SUF BPCs are constructed in an ISO 7 (Class 10,000) cleanroom, which complies with cGMP standards.
Features:
• Manufactured with industry-leading Aegis 5-14 film
• C-flex tubing that can be used with tubing welder and various connection options
• Configurable to meet your process needs, including options for single-use sensing (DO, pH, and pressure)
Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
Filters—used for bioburden reduction; sterilization grade filters and gas filters
Tubing—thermoplastic elastomers, platinum-cured silicone
Sample ports—septum-style and needle-free ports
Clamps—sliders, pinch, and crimp bands
Specifications
Product LineHyPerforma
Quantity30 L
Shipping ConditionRoom Temperature
Unit Size30 L
Contents & Storage
Store at room temperature.