Thermo Scientific™ Single-Use Fermentor BioProcess Container, CX5-14 film, traditional ports, ZenPure inlet, one 5-inch exhaust filter, 30 L
Thermo Scientific™
Thermo Scientific™ Single-Use Fermentor BioProcess Container, CX5-14 film, traditional ports, ZenPure inlet, one 5-inch exhaust filter, 30 L
Have Questions?
Catalog NumberQuantity
SH310390130 L, each
Catalog number SH3103901
Price (INR)
-
Quantity:
30 L, each
This 30 L Thermo Scientific Single-Use Fermentor (SUF) BioProcess Container (BPC) is composed of CX5-14 film, a five-layer, animal-origin free film, and includes traditional ports, ZenPure inlet, and one 5-inch exhaust filter.
All SUF BPCs are constructed in an ISO 7 (Class 10,000) cleanroom, which complies with cGMP standards.
Features:
• Manufactured with industry-leading CX5-14 film
• C-flex tubing that can be used with tubing welder and various connection options
• Configurable to meet your process needs, including options for single-use sensing (DO, pH, and pressure)
Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
Filters—used for bioburden reduction; sterilization grade filters and gas filters
Tubing—thermoplastic elastomers, platinum-cured silicone
Sample ports—septum-style and needle-free ports
Clamps—sliders, pinch, and crimp bands
Specifications
Product LineHyPerforma
Quantity30 L, each
Shipping ConditionRoom Temperature
Capacity (Metric)30 L
Unit Size30 L
Contents & Storage
Store at room temperature.