Labtainer™ 2D BioProcess Container (BPC), Aegis™ 5-14 film, three ports, Luer locks
Thermo Scientific™
Labtainer™ 2D BioProcess Container (BPC), Aegis™ 5-14 film, three ports, Luer locks
Thermo Scientific Labtainer 2D BioProcess Containers (BPCs) are composed of Aegis 5-14 single-web, multi-layer film.Features of the Labtainer 2D BPCRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
SH31050162000 mL
SH310501150 mL
SH3105012100 mL
SH3105013250 mL
SH3105014500 mL
SH31050151 L
SH31049015 L
SH310490210 L
SH310490320 L
SH310490450 L
SH31049052 L
Catalog number SH3105016
Price (INR)
-
Capacity (Metric):
2000 mL
Thermo Scientific Labtainer 2D BioProcess Containers (BPCs) are composed of Aegis 5-14 single-web, multi-layer film.
Features of the Labtainer 2D BPC include:
• Helps eliminate post-use cleaning steps required with re-useable containers and reduce cross-contamination risks
• Constructed in an ISO 7 cleanroom under cGMP conditions
Applications of the Labtainer 2D BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors
• Prepare and store buffer, culture media, and process liquid
• Collect chromatography feed and fractions
• Harvest, storage, and transport bulk drug product and bulk drug precursors
Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library with over 20,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters—used for bioburden reduction; sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands
Specifications
Capacity (Metric)2000 mL
Certifications/ComplianceISO 7
Film DescriptionAegis 5-14
For Use With (Application)Bioprocessing
Port Detail3 ports
Quantity2000 mL
Tubing StyleC-Flex
AutoclavableNot Autoclavable
ColorTransparent
DisposableSingle-use
SterilitySterile
Unit Size2 L