BioProcess Container (BPC), 50L, for HyPerforma™ Single-Use Mixer, Aegis™ 5-14 film, 3 ports
Thermo Scientific™
BioProcess Container (BPC), 50L, for HyPerforma™ Single-Use Mixer, Aegis™ 5-14 film, 3 ports
The 50L Thermo Scientific™ Bioprocess Containers (BPC), for powder-to-liquid mixing, is composed of CX5-14 single web, multi-layer plastic film andRead more
Have Questions?
Catalog NumberQuantity
SH31055.02
Catalog number SH31055.02
Price (INR)
-
The 50L Thermo Scientific™ Bioprocess Containers (BPC), for powder-to-liquid mixing, is composed of CX5-14 single web, multi-layer plastic film and includes 3 ports.
Single Use Mixer (S.U.M.) BPC options include:
• Open top liner for media/buffer hydration
• Closed BPC with powder port for contained media/buffer hydration
• Closed BPC with powder port and monitoring probe capability
• Closed BPC for liquid-liquid mixing of critical sterile solutions such as pooling of fractions or processing of intermediate solutions
• Closed BPC for liquid-liquid mixing with monitoring probe capability
Specifications
Film DescriptionCX5-14
For Use With (Application)Powder-Liquid BPC
Port Detail3 Probe Port
Product LineHyPerforma
Capacity (Metric)50 L