Harvestainer™ Microcarrier Separation System
Thermo Scientific™
Harvestainer™ Microcarrier Separation System
The Thermo Scientific™ Harvestainer Microcarrier Separation System is the ideal solution for the separation of microcarrier beads during adherent cell culture production.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
SH31078013 L
SH3107802
also known as SH31078.02
12 L
SH3107101
also known as SH31071.01
25 L
SH3107102
also known as SH31071.02
50 L
Catalog number SH3107801
Price (INR)
-
Capacity (Metric):
3 L
The Thermo Scientific™ Harvestainer Microcarrier Separation System is the ideal solution for the separation of microcarrier beads during adherent cell culture production. The Harvestainer system is a closed, single-use system that helps to increase product yields versus traditional methods, while reducing clean-in-place and steam-in-place requirements and water for injection usage. It eliminates equipment maintenance and reduces process validations, cycle times, and manual final decanting processes.
For smaller-scale or larger-scale applications, the Harvestainer system is ideal for in-line microcarrier separation. The 3 L and 12 L Harvestainer systems consist of a pre-assembled bioprocess container (BPC) and tray in a single-use unit, where the tray acts as the secondary containment device. The 25 L and 50 L Harvestainer systems consist of a 3D BPC with a drum as the secondary containment device. The BPC features a dip tube with dipwell design for better drainage and minimal manipulation.
  • Closed system
  • Scalable for both smaller-scale and larger-scale applications
  • Helps deliver high recovery rates
  • Inline microcarrier separation
Applications: Adherent cell culture production
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Capacity (Metric)3 L
MaterialPETG
Product LineHarvestainer
Quantity3 L
Shipping ConditionRoom Temperature
SterilitySterile
Unit Size3 L
Contents & Storage
Room Temperature