Product Image
Thermo Scientific™
Thermo Scientific™ 5:1 Single-Use Bioreactor BioProcess Container, CX5-14 film, drilled-hole sparger, crossflow sparger, 1000 L
Have Questions?
Catalog NumberQuantity
SH31132011,000 L
Catalog number SH3113201
Price (INR)
-
Quantity:
1,000 L
This 1000 L Thermo Scientific 5:1 Single-Use Fermentor (SUF) BioProcess Container (BPC) is composed of CX5-14 film, a five-layer, animal-origin free film, and includes a drilled-hole sparger (DHS) and crossflow sparger.
All SUF BPCs are constructed in an ISO 7 (Class 10,000) cleanroom, which complies with cGMP standards.
Features:
• Manufactured with industry-leading CX5-14 film
• C-flex tubing that can be used with tubing welder and various connection options
• Configurable to meet your process needs, including options for single-use sensing (DO, pH, and pressure)
Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
Filters—used for bioburden reduction; sterilization grade filters and gas filters
Tubing—thermoplastic elastomers, platinum-cured silicone
Sample ports—septum-style and needle-free ports
Clamps—sliders, pinch, and crimp bands
Specifications
Product LineHyPerforma
Quantity1,000 L
Capacity (Metric)1000 L
Unit Size1,000 L
Contents & Storage
Store at room temperature.