Transfer Assembly Y Manifold
Transfer Assembly Y Manifold
Transfer Assembly Y Manifold
Transfer Assembly Y Manifold
Thermo Scientific™
Transfer Assembly Y Manifold
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. Simplified connectivity and greater process efficiency for most upstream and downstream process operations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConnector TypesTubing Style
SH31487.01MPX Body, Y Connector, MPX Insert1/2 in. ID Braided Silicone
SH31200.01AseptiQuik S connectors/Y connector1/8 in. ID TPE
SH31458.01AseptiQuik G connectors/Y connector1/4 in. ID Silicone
SH31200.10AseptiQuik S connectors/Y connector1/4 in. ID TPE
SH31458.11AseptiQuik G connectors/Y connector3/8 in. ID Silicone
SH31200.20AseptiQuik S connectors/Y connector3/8 in. ID TPE
SH31458.21AseptiQuik G connectors/Y connector1/2 in. ID Silicone
SH31202.01AseptiQuik G connectors/Y connector1/4 in. ID TPE
SH31458.31AseptiQuik G connectors/Y connector3/4 in. ID Silicone
SH31202.11AseptiQuik G connectors/Y connector3/8 in. ID TPE
SH31461.01AseptiQuik S Connectors/Y Connector1/8 in. ID Silicone
SH31202.21AseptiQuik G connectors/Y connector1/2 in. ID TPE
SH31461.11AseptiQuik S Connectors/Y Connector1/4 in. ID Silicone
SH31202.31AseptiQuik G connectors/Y connector3/4 in. ID TPE
SH31461.21AseptiQuik S Connectors/Y Connector3/8 in. ID Silicone
SH31203.01AseptiQuik L connectors/Y connectors3/4 in. ID TPE
SH31203.11AseptiQuik L connectors/Y connector1 in. ID TPE
SH31470.01AseptiQuik L connectors/Y connector3/4 in. ID Silicone
SH31244.01MPX body, Y connector, MPX insert1/2 in. ID TPE
SH31471.01AseptiQuik L connectors/Y connectors1 in. ID Silicone
SH31246.01AseptiQuik L connectors/Y connectors1 in. ID TPE
SH31483.01MPC Body, Y Connector, MPC Insert3/8 in. ID Braided Silicone
SH31310.01MPC insert, Y connector, MPC body3/8 in. ID TPE
SH31485.01MPU Body, Y Connector, MPU Insert3/4 in. ID Silicone
SH31311.01MPU body, Y connector, MPU insert3/4 in. ID TPE
SH31486.01MPU Body, Y Connector, MPU Insert1 in. ID Braided Silicone
SH31247.01MPU Insert, Y Connector, MPU Body1 in. ID TPE
SH31312.01MPU Body, Y Connector, MPU Insert1 in. TPE
SH31472.01MPC Body, Y Connector, MPC Insert3/8 in. ID Silicone
SH31474.01MPU Insert, Y Connector, MPU Body1 in. ID Silicone
SH31475.01MPU Insert, Y Connector, MPU Body3/4 in. ID Silicone
SH31477.01MPX Body, Y Connector, MPX Insert1/2 in. ID Silicone
SH31478.01AseptiQuik G connectors/Y connector1/4 in. ID Braided Silicone
SH31478.11AseptiQuik G connectors/Y connector3/8 in. ID Braided Silicone
SH31478.21AseptiQuik G connectors/Y connector1/2 in. ID Braided Silicone
SH31478.31AseptiQuik G connectors/Y connector3/4 in. ID Braided Silicone
SH31479.11AseptiQuik S Connectors/Y Connector1/4 in. ID Braided Silicone
SH31479.21AseptiQuik S Connectors/Y Connector3/8 in. ID Braided Silicone
SH31480.01AseptiQuik L Connectors/Y Connector3/4 in. ID Braided Silicone
SH31481.11AseptiQuik L Connectors/Y Connector1 in. ID Braided Silicone
SH31482.01AseptiQuik L Connectors/Y Connectors1 in. ID Braided Silicone
Catalog number SH31487.01
Price (INR)
-
Connector Types:
MPX Body, Y Connector, MPX Insert
Tubing Style:
1/2 in. ID Braided Silicone
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. They connect to our standard Labtainer bioprocess containers (BPCs), Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. They connect to our standard Labtainer bioprocess containers (BPCs), Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
• One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
• Manufactured with BioTitan Retention Device—designed to provide superior universal tubing retention that minimizes the risk of leakage and failure at connection points
• Multiple manufacturing sites ensure security in product supply
• Produced under cGMP conditions in ISO 7 clean rooms to ensure a high level of quality
• Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard BPC products to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process. Designed to aid many common production processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
All Standard Fluid Transfer Assembly products are constructed of capped-end connectors and thermoplastic elastomer (TPE) tubing for use with welders. The Extension Set is available in a variety of tubing sizes and lengths to meet your fluid transfer needs.
These Standard Fluid Transfer Assembly products provide a common set of products that addresses an expansive set of needs. They are manufactured at multiple sites globally to minimize concerns of production interruptions and supply shortages. These products are built to our high quality standards to facilitate the success of your process.
Specifications
Connector TypesMPX Body, Y Connector, MPX Insert
DescriptionTransfer Assembly Y Manifold, 1/2 in. ID Braided Silicone, MPX Body, Y Connector, and MPX Insert
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Tubing Style1/2 in. ID Braided Silicone
Unit Size1 each
Contents & Storage
Room Temperature