InhibiSURE™ Viral Inactivation Medium
Thermo Scientific™
InhibiSURE™ Viral Inactivation Medium
Non-hazardous, inactivation media for viral collection and transport.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
SK01372Inactivation Medium NPG Swab Kit
EB1372AViral Inactivation Medium
Catalog number SK01372
Price (INR)
-
Product Type:
Inactivation Medium NPG Swab Kit
Rapidly inactivate and stabilize viral RNA at ambient temperature with a non-hazardous formulation using Thermo Scientific™ InhibiSURE™ Viral Inactivation Medium. This liquid medium is intended for the collection, transportation, and inactivation of SARS-CoV-2 and RNA enveloped viruses.
Collect and inactivate RNA enveloped viruses with Thermo Scientific™ InhibiSURE™ Viral Inactivation Medium. InhibiSURE stabilises viral RNA for transportation and use in subsequent in vitro diagnostic testing procedures.  The device is intended to be used with unprocessed nasal swabs, nasopharyngeal swabs and throat swabs.  This is a single use device, it is not automated nor is it a companion diagnostic.
Product is CE marked but not 510(k)-cleared and not available for sale in the U.S. Availability of product in each country depends on local regulatory marketing authorization status. - For In Vitro Diagnostic Use.
For In Vitro Diagnostic Use.  
Specifications
Closure ColorPurple
CompatibilityInhibiSURE™ Viral Inactivation Medium has been designed to be compatible with nucleic acid purification and amplification workflows. It has been validated with the Thermo Scientific™ MagMax™ MVP II kit (product code A48383) and TaqPath™ COVID 19 CE IVD RT PCR Kit (A48067)
DescriptionInhibiSURE™ Viral Inactivation Medium NPG Swab Kit
Fill Volume1.5 mL
For Use With (Application)Medium is a liquid medium intended to be used for the collection and inactivation of RNA enveloped viruses. The product stabilises the viral RNA for transportation and use in subsequent in vitro diagnostic testing procedures. The device is intended to be used with unprocessed nasopharyngeal swabs.
Quantity100
Storage Requirements 2 - 30°C
Transport Temperature2 - 30°C
Product TypeInactivation Medium NPG Swab Kit
Unit SizeEach