Sensititre™ SLOMYCO2 Susceptibility Testing Plate
Thermo Scientific™
Sensititre™ SLOMYCO2 Susceptibility Testing Plate
Updated susceptibility testing for slow growing mycobacterium which now includes Clofazimine, with high-quality, easy-to-use broth microdilution (BMD) plates, designed with reference to the latest CLSI guidelines (Research Use Only)
Have Questions?
Catalog number SLOMYCO2
Price (INR)
-
Perform susceptibility testing for slow growing mycobacterium species (SGM) using broth microdilution, the recommended method for Antimicrobial Susceptibility Testing. The Thermo Scientific™ Sensititre™ SLOMYCO2 plate offers scientists an updated (from SLOMYCO) plate format that aligns with recently updated guidelines for susceptibility testing of nontuberculous mycobacteria.
Gain MIC data for an extended range of relevant antibiotics. This updated format includes the addition of Clofazimine, an option now considered in multi-drug regimens associated with macrolide resistant SGM isolates.
Achieve reliable, consistent results for slowly growing non-tuberculosis mycobacteria isolates with Sensititre SLOMYCO2 AST Plate.
  • Broth microdiltion (BMD) is the recommended AST method for mycobacteria
  • Test Mycobacterium spp. including slowly growing Mycobacterium avium group, M. kansasii group or M. marinum group.
  • Designed with reference to the latest CLSI advice on drug options and accompanying breakpoints (where set)
  • BMD is a recommended AST method that maintains accurate results even where other methodologies fail
  • Extended shelf life, room-temperature storage eliminates inventory control concerns
  • Individual packaging allows the laboratory to test one plate at a time with no waste
  • Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology
  • Easy set-up procedures and results in approximately 14 days for slow-growing organisms
  • Test 12 antimicrobials on a single AST plate for cost-effective results
Thermo Fisher Scientific is a leader in providing AST solutions that help in the fight against antimicrobial resistance, offering high performance automated and manual test options and one of the largest, most up-to-date selections of antimicrobials for susceptibility testing.
The Sensititre System utilizes gold standard-level MIC accuracy1, providing superior quality and reproducibility for accurate results the first time. Broth microdilution is the reference method for AST, and in many cases 2 the most reliable phenotypic method to guide therapeutic decisions.
For Research Use Only. Not for use in diagnostic procedures.
General References:
  1. International Organization for Standardization (ISO) (2019) Susceptibility testing of infectious agents and evaluation of performance of antimicrobial susceptibility test devices — Part 1: Broth micro-dilution reference method for testing the in vitro activity of antimicrobial agents against rapidly growing aerobic bacteria involved in infectious diseases. ISO 20776-1:2019 2. EUCAST (2019) EUCAST reading guide for broth microdilution. Version 1. Accessed Nov 2019
  2. http://www.eucast.org/fileadmin/src/media/PDFs/EUCAST_files/Disk_test_documents/2019_manuals/Reading_guide_BMD_v_1.0_2019.pdf ...for antimicrobial susceptibility testing of rapidly growing aerobic bacteria, except for mecillinam and fosfomycin, where agar dilution is the reference method.
Specifications
Antibiotic CodeVarious
Antimicrobial AgentsVarious
DescriptionSensititre Myco Susceptibility Plate – Slow growing mycobacterium
No. per Pack10
Packaging TypeBox
Quantity10/Pk.
Product TypeMIC Plate
Target Organism ClassSlow-growing Mycobacteria
Test TypeMinimum Inhibitory Concentration (MIC)
Unit SizeEach