Product Image
Thermo Scientific™

Sensititre™ SLOMYCOI

Confidently perform susceptibility testing of slow-growing non-tuberculosis mycobacteria isolates with this convenient and cost-effective plate.

Have Questions?
Catalog NumberQuantity
SLOMYCOI10/Pk.
Catalog number SLOMYCOI
Price (INR)
-
Quantity:
10/Pk.

Confidently perform susceptibility testing of slow-growing non-tuberculosis mycobacteria isolates with Thermo Scientific™ Sensititre™ Myco SLOMYCOI AST Plate. This standard, off-the-shelf, AST plate format includes 13 antimicrobials for cost effective AST testing, and is intended for In Vitro Diagnostic Use.

Obtain true minimum inhibitory concentration (MIC) results to inform targeted therapy decisions and to support antimicrobial stewardship to preserve effectiveness.

Achieve reliable, consistent results for slow-growing non-tuberculosis mycobacteria isolates with Sensititre Myco SLOMYCOI AST Plate. Extended shelf life, room-temperature storage eliminates inventory control concerns

  • Individual packaging allows the laboratory to test one plate at a time with no waste
  • Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology
  • Easy set-up procedures and results up to 14 days for slow growing organisms.
  • Convenient microbroth dilution format provides accurate endpoint determination with manual read or Thermo Scientific™ Sensititre™ Vizion™ System options

The Sensititre System is a leader in AST solutions, offering flexible, customizable testing options for laboratories of all sizes.

  • Scalable – flexible instrumentation options designed to be modular in order to meet the unique needs of individual lab space, workload, and budget requirements
  • Flexible – over 300 antimicrobials available in extended dilution ranges, access to the newest antimicrobials, and the ability to consolidate AST onto one platform
  • Tailor-made – design your own AST panel to meet specific formulary requirements and prescription protocols based on real-time trends and local epidemiology

Not all products are available for sale in all territories. Please inquire.

VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.

For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
0.5 McFarland StandardH2O (Part No. T3339)
Antibiotic CodeVarious
Antimicrobial AgentsVarious
Broth TypeMHB w/ OADC (Part No. T8005)
DescriptionSensititre Myco Susceptibility Plate- Slow Growing
For Use With (Application)Slow-growing Mycobacteria
Incubation Conditions35°C O2
Incubation Time7-21 days
Isolates Per Plate1
No. per Pack10
Packaging TypeBox
Quantity10/Pk.
Reading OptionsManual
Transfer Volume50µL
Volume Per Well100 μL
Product TypeMIC Plate
Target Organism ClassRapidly-growing, Non-tuberculous Mycobacteria
Test TypeMicrobial Susceptibility
Unit SizeEach