Oxoid™ OGYE Selective Supplement
Thermo Scientific™
Oxoid™ OGYE Selective Supplement
Oxoid OGYE Selective Supplement (Oxytetracycline Glucose Yeast Extract) is added to Oxytetracycline Glucose Yeast Extract Agar Base, Part No. CM0545B.
Have Questions?
Catalog NumberQuantity
SR0073A10/Pk.
Catalog number SR0073A
Price (INR)
-
Quantity:
10/Pk.
Oxoid OGYE Selective Supplement (Oxytetracycline Glucose Yeast Extract) is added to Oxytetracycline Glucose Yeast Extract Agar Base, Part No. CM0545B.
  • Add to Oxytetracycline Glucose Yeast Extract Agar Base, Part No. CM0545B
  • Each vial supplements 500mL of medium
Specifications
DescriptionOGYE Selective Supplement
Quantity10/Pk.
YieldEach vial for 500mL medium
FormFreeze-dried
Product TypeSelective Supplement
Unit SizeEach