Oxoid™ Dermasel Selective Supplement
Thermo Scientific™
Oxoid™ Dermasel Selective Supplement
The Oxoid Dermasel Selective Supplement is used for Dermasel Medium, Part No. CM0539B.
Have Questions?
Catalog NumberQuantity
SR0075E10/Pk.
Catalog number SR0075E
Price (INR)
-
Quantity:
10/Pk.
Oxoid Dermasel Selective Supplement is used for Dermasel Medium, Part No. CM0539B.
Each vial supplements 500mL of medium
Specifications
DescriptionDermasel Selctv Supp.
Quantity10/Pk.
YieldEach vial for 500mL medium
FormFreeze-dried
Product TypeSelective Supplement
Unit SizeEach