Oxoid™ Bordetella Selective Supplement
Thermo Scientific™
Oxoid™ Bordetella Selective Supplement
Oxoid Bordetella Selective Supplement (Cephalexin, 20mg/vial) isolates Bordetella pertussis and B. parapertussis.
Have Questions?
Catalog NumberQuantity
SR0082E10/Pk.
Catalog number SR0082E
Price (INR)
-
Quantity:
10/Pk.
Oxoid Bordetella Selective Supplement (Cephalexin, 20mg/vial) is used for the selective isolation of Bordetella pertussis and B. parapertussis.
Add to to Charcoal Agar Base, Part No. CM0119B. Each vial supplements 500mL of medium.
Specifications
DescriptionBordetella Selective Supplement
Quantity10/Pk.
YieldEach vial for 500mL medium
FormFreeze-dried
Product TypeSelective Supplement
SpeciesBordetella species
Unit SizeEach