Oxoid™ Vitox Supplement
Thermo Scientific™

Oxoid™ Vitox Supplement

Oxoid Vitox Supplement is used for the preparation of Thayer Martin Medium and New York City Medium for the isolation of Neisseria spp.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberYield
SR0090AEach vial for 500mL medium
SR0090HEach vial for 2L medium
Catalog number SR0090A
Price (INR)
-
Yield:
Each vial for 500mL medium

Oxoid Vitox Supplement is used for the preparation of Thayer Martin Medium and New York City Medium for the isolation of Neisseria spp.

  • Contents: Vitox Supplement (5 vials) plus reconstitution fluid (5 vials)
  • Add to GC Agar Base, Part No. CM0367B

Ordering Information:

  • Each vial of Vitox plus reconstitution fluid supplements 500mL of medium

Includes:

  • Five vials of Vitox plus five vials of reconstitution fluid
Specifications
DescriptionVitox Supplement
Packaging Type2 x 5/Pk.
Quantity5 x Freeze dried vial + 5 x Rehydration fluid
YieldEach vial for 500mL medium
FormFreeze-dried
Product TypeMicrobiology Media
Unit SizeEach