Oxoid™ VCNT Selective Supplement
Product Image
Thermo Scientific™
Oxoid™ VCNT Selective Supplement
The Oxoid VCNT Selective Supplement is used for the preparation of Thayer Martin Medium for the isolation of pathogenic Neisseria spp.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberYield
SR0091EEach vial for 500mL medium
SR0091HEach vial for 2L medium
Catalog number SR0091E
Price (INR)
-
Request A Quote
Yield:
Each vial for 500mL medium
Oxoid VCNT Selective Supplement is used for the preparation of Thayer Martin Medium for the isolation of pathogenic Neisseria spp.
  • Add to GC Agar Base, Part No. CM0367B
Specifications
DescriptionVCNT Selective Supplement
Quantity10/Pk.
YieldEach vial for 500mL medium
FormFreeze-dried
Product TypeSelective Supplement
Unit SizeEach