Product Image
Thermo Scientific™
Oxoid™ N-S Anaerobe Selective Supplement
The Oxoid N-S Anaerobe Selective Supplement is used for the growth of fastidious and other anaerobes.
Have Questions?
Catalog NumberQuantity
SR0107B10/Pk.
Catalog number SR0107B
Price (INR)
-
Quantity:
10/Pk.
Oxoid N-S Anaerobe Selective Supplement is used for the growth of fastidious and other anaerobes.
  • Add to Wilkins-Chalgren Anaerobe Agar, Part No. CM0619B
Specifications
DescriptionN-S Anaerobe Selective Supplement
Quantity10/Pk.
YieldEach vial for 500mL medium
FormFreeze-dried
Product TypeSelective Supplement
SpeciesAnaerobes
Unit SizeEach