Oxoid™ Broad Spectrum Mixture Beta Lactamase
Thermo Scientific™
Oxoid™ Broad Spectrum Mixture Beta Lactamase
Mixture of beta-lactamase enzymes from Bacillus cereus for the inactivation of beta-lactam antibiotic preparations present in specimens.
Have Questions?
Catalog NumberQuantity
SR0113E10/Pk.
Catalog number SR0113E
Price (INR)
-
Quantity:
10/Pk.
Oxoid Beta Lactamase, Broad Spectrum Mixture is a mixture of beta-lactamase enzymes from Bacillus cereus for the inactivation of beta-lactam antibiotic preparations present in specimens.
Specifications
DescriptionBeta Lactamase, Broad Spectrum Mixture
Quantity10/Pk.
FormFreeze-dried
Product TypeBeta Lactamase Mixture
Unit SizeEach