Product Image
Thermo Scientific™
Oxoid™ RPF Supplement
The Oxoid RPF Supplement (Rabbit Plasma Fibrinogen) is used for isolation and enumeration of coagulase positive staphylococci from food.
Have Questions?
Catalog NumberQuantity
SR0122A10/Pk.
Catalog number SR0122A
Price (INR)
-
Quantity:
10/Pk.
Oxoid RPF Supplement (Rabbit Plasma Fibrinogen) is used for the isolation and enumeration of coagulase positive staphylococci from food.
  • Use only with Baird-Parker Agar Base, RPF, Part No. CM0961B
  • Each vial supplements 100mL of medium
Specifications
DescriptionRPF Supplement
Quantity10/Pk.
FormFreeze-dried
Product TypeMicrobiology Media
Unit SizeEach