Product Image
Oxoid UK

Oxoid™ Penase 1 vial

A lactamase enzyme from Bacillus cereus for the inactivation of a range of penicillins.
Have Questions?
Catalog number SR0129B
Price (INR)
-
A Bacillus cereus 569/H9 lactamase (E.C.5.2.6) presented as a freeze-dried powder containing buffer salts. Each vial contains a 3,300lU of activity (1 unit of enzyme activity will hydrolyse 1.0μmol of benzylpenicilloic acid per minute pH7.0 and at 25°C). Used for the inactivation of susceptible beta-lacram antibiotics in preparations, blood or tissue samples.
Specifications
Country of OriginUK
For Use With (Application)Inactivation of a Range of Penicillins
FormatVial
Media TypeSupplement
Packaging TypeVial
Quantity1 Each
Shelf Life12 months
Product TypeMedia Supplement
Unit SizeEach
Contents & Storage
Store at 2-8°C.