Oxoid™ UVM1 Listeria Primary Selective Enrichment Supplement
Thermo Scientific™
Oxoid™ UVM1 Listeria Primary Selective Enrichment Supplement
The Thermo Scientific™ Oxoid Listeria Primary Selective Enrichment Supplement, UVM1 is a selective method for enrichment of Listeria spp.
Have Questions?
Catalog NumberDescription
SR0142EListeria Primary Selective Enrichment Supplement, UVM1
Catalog number SR0142E
Price (INR)
-
Description:
Listeria Primary Selective Enrichment Supplement, UVM1
Thermo Scientific™ Oxoid Listeria Primary Selective Enrichment Supplement, UVM1 is used in a two-step selective enrichment (USDA-FSIS) method for the enrichment of Listeria spp.
Add to Listeria Enrichment Broth Base (UVM formulation), Part No. CM0863B
Specifications
DescriptionListeria Primary Selective Enrichment Supplement, UVM1
Quantity10/Pk.
YieldEach vial for 500mL medium
FormFreeze-dried
Product TypeSelective Supplement
SpeciesListeria
Unit SizeEach