Oxoid™ TTC Solution
Thermo Scientific™

Oxoid™ TTC Solution

Oxoid TTC Solution (0.125%) is used for the enumeration and identification of coliform bacteria.
Have Questions?
Catalog NumberQuantity
SR0148A10 x 2 mL
Catalog number SR0148A
Price (INR)
-
Quantity:
10 x 2 mL

Oxoid TTC Solution (0.125%) is used for the enumeration and identification of coliform bacteria.

Add to Tergitol 7 Agar, Part No. CM0793B
Specifications
DescriptionTTC Solution
Quantity10 x 2 mL
YieldEach vial for 100mL medium
FormLiquid
Product TypeSolution
Unit SizeEach