Oxoid™ Modified Listeria Selective Enrichment Supplement
Thermo Scientific™
Oxoid™ Modified Listeria Selective Enrichment Supplement
Oxoid Listeria Selective Enrichment Supplement, Modified is used for the enrichment of Listeria spp.
Have Questions?
Catalog NumberQuantity
SR0149A10/Pk.
Catalog number SR0149A
Price (INR)
-
Quantity:
10/Pk.
Oxoid Listeria Selective Enrichment Supplement, Modified is used for the enrichment of Listeria spp.
  • Modified with 10mg/L acriflavin
  • Add to Listeria Enrichment Broth, Part No. CM0862B
  • Each vial supplements 2.25L of medium
Specifications
DescriptionListeria Selective Enrichment Supplement, Modified
Quantity10/Pk.
YieldEach vial for 2.25L medium
FormFreeze-dried
Product TypeSelective Supplement
SpeciesListeria
Unit SizeEach