Product Image
Thermo Scientific™
Oxoid™ STAA Selective Supplement
The Oxoid STAA Selective Supplement is used for the isolation of Brochothrix thermosphacta from food samples.
Have Questions?
Catalog NumberQuantity
SR0151E10/Pk.
Catalog number SR0151E
Price (INR)
-
Quantity:
10/Pk.
Oxoid STAA Selective Supplement is used for the isolation of Brochothrix thermosphacta from food samples.
  • Add to STAA Agar Base, Part No. CM0881B
  • Each vial supplements 500mL of medium
Specifications
DescriptionSTAA Selective Supplement
Quantity10/Pk.
YieldEach vial for 500mL medium
FormFreeze-dried
Product TypeSelective Supplement
Unit SizeEach