Oxoid™ CCDA Selective Supplement
Thermo Scientific™

Oxoid™ CCDA Selective Supplement

Oxoid Brain Heart Infusion Broth is highly nutritious medium for the growth of fastidious organisms, recommended for blood culture work; also see Heart Infusion Broth.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityYield
SR0155E10/Pk.Each vial for 500mL medium
SR0155H10 VialsFor 2L medium
Catalog number SR0155E
Price (INR)
-
Quantity:
10/Pk.
Yield:
Each vial for 500mL medium

Oxoid CCDA Selective Supplement is used for the isolation of Campylobacter spp., especially C. upsaliensis or Arcobacter.

  • For the isolation of Arcobacter spp., add to Arcobacter Broth Base, Part No. CM0965B
  • For the isolation of Campylobacter spp., especially C. upsaliensis, add to Campylobacter Blood-Free Selective Agar Base (CCDA Preston), Part No. CM0739B
Specifications
DescriptionCCDA Selective Supplement
Quantity10/Pk.
YieldEach vial for 500mL medium
FormFreeze-dried
Product TypeSelective Supplement
Unit SizeEach