Product Image
Thermo Scientific™
Oxoid™ STA Selective Supplement
The Oxoid STA Selective Supplement (Supplement without actidione) is used for the isolation of Brochothrix thermosphacta from food samples.
Have Questions?
Catalog NumberQuantity
SR0162E10/Pk.
Catalog number SR0162E
Price (INR)
-
Quantity:
10/Pk.
Oxoid STA Selective Supplement (Supplement without actidione) is used for the isolation of Brochothrix thermosphacta from food samples.
  • Add to STAA Agar Base, Part No. CM0881B
  • Each vial supplements 500mL of medium
Specifications
DescriptionSTA Selective Supplement
Quantity10/Pk.
YieldEach vial for 500mL medium
FormFreeze-dried
Product TypeSelective Supplement
Unit SizeEach