Oxoid™ Neomycin Selective Supplement
Thermo Scientific™
Oxoid™ Neomycin Selective Supplement
The Oxoid Neomycin Selective Supplement (150mg/vial) allows the growth of clostridia and other anaerobes.
Have Questions?
Catalog NumberQuantity
SR0163H10 x 150 mg
Catalog number SR0163H
Price (INR)
-
Quantity:
10 x 150 mg
Oxoid Neomycin Selective Supplement (150mg/vial) allows the growth of clostridia and other anaerobes.
  • Add to Blood Agar, Wilkins-Chalgren Anaerobe Agar, Part No. CM0619B, or Columbia Blood Agar, Part No. CM0331B
  • Each vial supplements 2L of medium
Specifications
DescriptionNeomycin Selective Supplement
Quantity10 x 150 mg
YieldEach vial for 2L medium
FormFreeze-dried
Product TypeSelective Supplement
Unit SizeEach