Oxoid™ Karmali Campylobacter Selective Supplement
Thermo Scientific™
Oxoid™ Karmali Campylobacter Selective Supplement
Oxoid Campylobacter Selective Supplement, Karmali isolates Campylobacter jejuni and C. coli.
Have Questions?
Catalog NumberQuantity
SR0167E10/Pk.
Catalog number SR0167E
Price (INR)
-
Quantity:
10/Pk.
Oxoid Campylobacter Selective Supplement, Karmali is used for the isolation of Campylobacter jejuni and Campylobacter coli when incubated at 42°C.
  • Add to Campylobacter Agar Base (Karmali), Part No. CM0935B
  • Each vial supplements 500mL of medium
Specifications
DescriptionCampylobacter Selective Supplement, Karmali
Quantity10/Pk.
YieldEach vial for 500mL medium
FormFreeze-dried
Product TypeSelective Supplement
SpeciesCampylobacter
Unit SizeEach