Oxoid™ CAT Supplement
Actual product may vary
Thermo Scientific™
Oxoid™ CAT Supplement
The Oxoid CAT Supplement isolates Campylobacter spp.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
SR0174E10/Pk.
SR0174HFor 2L medium
Catalog number SR0174E
Price (INR)
-
Quantity:
10/Pk.
Oxoid CAT Supplement is used for the isolation of Campylobacter spp., especially C. upsaliensis or Acrobacter.
  • Add to Campylobacter Blood-Free Selective Agar Base (CCDA Preston), Part No. CM0739B for the isolation of Campylobacter spp.
  • Add to Arcobacter Broth Base, Part No. CM0965B for the isolation of Arcobacter spp.
Specifications
DescriptionCAT Supplement
Quantity10/Pk.
YieldEach vial for 500mL medium
FormFreeze-dried
Product TypeMicrobiology Media
SpeciesCampylobacter
Unit SizeEach