Oxoid™ Meropenem Selective Supplement
Thermo Scientific™
Oxoid™ Meropenem Selective Supplement
Used for the isolation of Vancomycin-Resistant Enterococci (VRE) and HighLevel Aminoglycoside-Resistant Enterococci (HLARE) from clinical specimens.
Have Questions?
Catalog NumberQuantity
SR0184E10/Pk.
Catalog number SR0184E
Price (INR)
-
Quantity:
10/Pk.
Oxoid Meropenem Selective Supplement (1mg/vial) is used for the isolation of Vancomycin-Resistant Enterococci (VRE) and HighLevel Aminoglycoside-Resistant Enterococci (HLARE) from clinical specimens.
  • Add to VRE Agar Base, Part No. CM0984B
  • Each vial supplements 500mL of medium
Specifications
DescriptionMeropenem Selective Supplement
Quantity10/Pk.
YieldEach vial for 500mL medium
FormFreeze-dried
Product TypeSelective Supplement
Unit SizeEach