Oxoid™ Vancomycin Supplement
Thermo Scientific™
Oxoid™ Vancomycin Supplement
Used for addition to selective media for isolation of Vancomycin Resistant Enterococci and High Level Aminoglycoside Resistant Enterococci from clinical specimens.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormat
SR0186E10/Pk.
SR0247E10/Pk.
Catalog number SR0186E
Price (INR)
-
Quantity:
10/Pk.
Thermo Scientific™ Oxoid Vancomycin Supplement is for addition to selective media for the isolation of Vancomycin Resistant Enterococci (VRE) and High Level Aminoglycoside Resistant Enterococci (HLARE) from clinical specimens.
  • Each vial supplements 500mL of medium
Specifications
DescriptionOxoid™ Vancomycin Supplement
Quantity10/Pk.
FormFreeze-dried
Product TypeMicrobiology Media
Unit SizeEach