A chromogenic medium for rapid and presumptive identification of Clostridium perfringens from water samples.
Have Questions?
Catalog Number
Quantity
SR0188E
10/Pk.
Catalog number SR0188E
Price (INR)
-
Quantity:
10/Pk.
Oxoid Membrane Clostridium Perfringens Selective Supplement (m-CP) is a chromogenic medium for rapid and presumptive identification of Clostridium perfringens from water samples.
Add to Membrane Clostridium Perfringens (m-CP) Agar Base, Part No. CM0992B