Product Image
Thermo Scientific™
Oxoid™ Modified Karmali Selective Supplement
A blood-free selective medium for the isolation of Campylobacter jejuni and Campylobacter coli when incubated at 42° C.
Have Questions?
Catalog NumberQuantity
SR0205E10/Pk.
Catalog number SR0205E
Price (INR)
-
Quantity:
10/Pk.
Oxoid Modified Karmali Selective Supplement is for a blood-free selective medium for the isolation of Campylobacter jejuni and Campylobacter coli when incubated at 42° C.
  • Alternative supplement containing amphotericin B as a replacement for cycloheximide
  • Add to Campylobacter Agar Base (Karmali), Part No. CM0935B
  • Each vial supplements 500mL of medium
Specifications
DescriptionModified Karmali Selective Supplement
Quantity10/Pk.
YieldEach vial for 500mL medium
FormFreeze-dried
Product TypeSelective Supplement
SpeciesKarmali
Unit SizeEach